Autosampler does not reach set temperature
Possible Cause
Remedial Action
Ambient temperature or humidity too high
  1. Check that the insulation cover is installed.
  2. Reduce the ambient temperature (for example, by ventilating the room), and/or ambient humidity.