Column compartment temperature does not change although the set point has not been reached
Possible Cause | Remedial Action |
---|---|
Ambient temperature too high or setpoint lower than the specified difference to the ambient temperature |
|
Temperature control turned off | Turn on the temperature control. |
Ventilation opening obstructed |
|
Ambient air enters column chamber | Improper sealing of the column chamber may impair the heating performance of the column compartment.
|
Column compartment defective | Contact Technical Support. |