Product component | What you should do... |
---|---|
System/all modules | Have Thermo Fisher Scientific service personnel perform preventive maintenance once a year. |
Seal wash system |
|
Pump | Inspect the pistons for particle deposits or indications of damage, for example, scratches or scores. See Cleaning the Piston. |