Product component
What you should do...
System/all modules
  • Flow connections:
    Inspect flow connections for signs of:
    • Blockage: Run the script and follow the instructions: D01 - Test System Back Pressure.
    • Salt deposits
    • Air bubbles
      Sonicate the solvents and purge the pump and the autosampler if necessary.
  • Solvents:
    If you use one of the following solvents, proceed as follows:
    • Pure water or an aqueous buffer: To replace the solvent reservoir, run the script: B01 - Change Liquids / Solvents.
    • Buffers or salt solutions: Flush the system thoroughly after use with an appropriate solvent that does not contain buffers or salts. See Purging (Overview).
    • Corrosive solvents or methanol with <10% water: Flush the system with water or other suitable non-corrosive solvents. See Purging (Overview).
Wash systems
  • Check the liquid level.
  • Check if particles, dust, or algae are present in the wash reservoirs.
    If particles, dust, or algae are present, replace the wash reservoirs with clean ones. Run the script: B01 - Change Liquids / Solvents (Refresh Only = On). Ensure that the wash reservoirs are free of detergents or residual chemicals. We recommend rinsing the wash reservoirs with a small quantity of the planned wash liquid.
Autosampler
  • Inspect vials and well plates in the autosampler for cracks or defects.
  • Clean up spills inside the autosampler if necessary.