The following connectors are provided on the short panel of the system controller:
No. | Description |
|---|---|
1 | Rating plate Indicates the serial number and the line rating. |
2 | Vanquish System Controller (VSC) connector Allows connection to the system base. |
3 | System Interlink port Allows connection to the system base and to a module in the HPLC system |
4 | Universal Serial Bus (USB) port ("A" type connector) Deactivated – Do not use |
5 | Local Area Network (LAN) port Allows connection to the site network. |
6 | Main power switch (on/off control) |
7 | Power inlet port |
The following connectors and slots are provided on the long panel of the system controller:
No. | Description |
|---|---|
1 | Display power outlet port |
2 | Display data port |
3 | System Interlink ports Deactivated |
4 | USB port ("A" type connector) Deactivated - Do not use |
5 | Label indicating the part number, module name and revision number. |
6 | Storage Device (SD) card slot |