The user-accessible components of the pump are located directly behind the front doors.
1234356789111213a13b13c13d13xxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxxx10ABAB
Figure: Pump interior components
No.
Description
1
Keypad with buttons for pump control
2
Pump head A (left pump head)
3
Tubing clips for routing solvent lines to the pump heads
4
Pump head B (right pump head)
5
Shut-off valves
6
Leak sensor
7
Type label, indicating the module name, serial number, part number, and revision number
8
Drain port
9
Seal wash detector (drop detector)
10
Degasser for autosampler needle wash liquids
11
Seal wash pump
12
Tubing chase with tubing guides
13
Flowmeter, with:
13a
Flowmeter inlet B
13b
Pump outlet (flowmeter outlet)
13c
Flowmeter waste line (Purge Out)
13d
Flowmeter inlet A