Parts required
Bottle insert
Follow these steps
  1. Insert the rear and the left wing into the gap below the railings.
  2. Firmly push the right wing below the railing.
  3. 112
    Figure: Inserting the bottle insert
  4. Place the liquid reservoirs into the solvent rack bottle insert.
  5. Connect the liquid lines to the liquid reservoirs.
  6. Turn on the system and the system controller with the power buttons on the left side of the system base.
  7. Prepare the system for initial operation. See Preparing the System for Initial Operation.