Parts required
Lint-free tissue
Follow these steps
  1. Turn off the system with the system power button (lower button on the system base).
  2. Turn off the system controller with its power button (upper button on the system base).
  1. Disconnect the seal wash lines:
  1. Unscrew the cap of the seal wash reservoir.
  2. Together with the cap, remove the seal wash line from the seal wash reservoir.
  1. Disconnect the solvent lines:
  1. Unscrew the caps of the solvent reservoirs.
  2. Together with the caps, remove the solvent lines from the reservoirs.
  3. Protect the solvent line filters from contamination as appropriate.
  1. Absorb any liquid from the liquid lines with a lint-free tissue.
  2. Remove the liquid reservoirs from the solvent rack.
  3. Firmly push the bottle insert wings above the solvent rack railing and remove the bottle insert.
111
Figure: Removing the bottle insert
No.
Description
1
Wing