CAUTION
Heavy Load, Bulky System
The system is too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the system, observe the following guidelines:
  • Physical handling of the system, including lifting or moving, requires a team effort of two persons.
  • Use the carrying handles that are fixed to the system to move or transport the system. Never move or lift the system by the front doors. This will damage the doors or the system modules. Verify that the handles are fixed properly and cannot slide off the system.
NOTICE
Moving the Vanquish system stack even over short distances on the workbench can damage and impair the functionality of the system base.
Before moving the system stack, unlock the system base using the locking tools.
Tools required
Screwdriver, Torxâ„¢ T30
Follow these steps
  1. Transport the system to the installation site.
  2. Remove all straps from the shipping container.
  3. Lift the cardboard cover and remove it.
  4. Remove the ship kit.
  5. Lift the upper foam piece and remove it.
  6. Remove the system from the shipping container:
  1. Grasp the system by the carrying handles.
  2. Slowly and carefully, lift the system out of the shipping container.
  1. Place the system on a stable surface and leave protective films attached to the surfaces.
  2. To be able to shift the system base slightly on the workbench for better access to system parts, unlock the system base using the system base locking tools. See Unlocking the System Base.
  3. Remove any additional packing material.
  4. Optional: Remove the carrying handles:
  1. On each carrying handle, loosen the attachment screws until the carrying handles are moveable in the rail. Do not remove the screws from the carrying handles completely.
  2. Slide off the carrying handles from the rails toward the rear of the system.
Figure: Sliding off the carrying handles from the left rail
  1. Fasten the screws of the carrying handles.
  2. TIP
    Keep the shipping container, the carrying handles with the attachment screws, and all packing material. These items will be needed if the system is transported to a new location.
  3. Remove the protective film from all surfaces as applicable.