The system base has a locking mechanism. Before you move the system base, for example, for installation and maintenance, you need to unlock the system base. To prevent it from being moved during operation on the workbench, lock it.
You can switch between the moveable and stationary mode using the system base locking tools shipped with the system base.
System base locking tool
Figure: System base locking tool