The system base provides:
  • Power buttons for power on/off control of all modules and the system controller
  • Drawer to store tools and small system parts
  • Drain port for connecting a system waste line
  • Locks to toggle between moveable and stationary mode
System base (front and rear view)
Figure: System base (front and rear view)
No.
Description
1
Vanquish System Controller power button
Allows power on/off control of the system controller.
2
System power button
Allows power on/off control for all modules of the system that are connected to the system base via the System Interlink port.
3
Drawer for tools
Allows storing any tools that are necessary to install or maintain the Vanquish system. Push the drawer front to open or close the drawer.
4
Reserved for future use
5
System drain port
For connecting the system waste line.
6
Locks - on the left and right side of the system base
Allows toggling between movable and stationary mode.
7
System Interlink port
Allows connecting the other modules for power on/off control from the Vanquish system base.
8
VSC port
Allows connecting the system controller.