Follow the steps below to replace the wash line between outlet of the metering device head and the inlet of the seal wash pump.
When
Seal was pump inlet line is clogged or damaged
Parts required
Silicone tubing and connectors
Tools required
Tubing cutter
Preparations
Empty the seal wash system:
- Unscrew the cap of the seal wash reservoir.
- Together with the cap, remove the seal wash line from the seal wash reservoir.
- To empty the seal wash line, lift up the lever of the seal wash pump. Release the lever when the line is empty.
Follow these steps
Pull the tubing connector with the seal wash pump inlet line out of the seal wash outlet port on the metering device head.
Disconnect the seal wash pump inlet line from the peristaltic tubing on the seal wash pump in the pump.
Connect a new tubing connector to the new seal wash pump inlet line. Cut tubing to required length, if applicable.
Connect the new seal wash pump inlet line to the seal wash outlet on the metering device head.
Connect the seal wash pump inlet line to the peristaltic tubing on the seal wash pump.
Restarting the Seal Wash System
Retighten the reservoir cap hand-tight. The retaining guide must remain in the hole in the reservoir cap. If it does not, press the retaining guide into the hole so that the tubing is kept in place in the cap.
Check the wash lines over the entire flow path:
- Make sure that no bending (kink), pinching or squeezing of the wash lines is present at any point in the flow path.
- If you have to cut tubing to length, use a tubing cutter. Make sure that the cut is at right angle to the length of the line.
Flush the seal wash system. Run the script: B01 - Change Liquids / Solvents (with: Rear Seal Wash Liquid and Refresh Only = Off.)