In the direct injection setup the injection valve ports are assigned as follows:
Port | Connected Component |
|---|---|
1 | Viper plug |
2 | Solvent valve |
3 | Needle seat |
4 | Separation Column (if installed: Column Compartment) |
5 | Sample loop |
6 | Metering device |
7 | Pump |
For direct injection and trap-and-elute workflows, the solvent valve ports are assigned as follows:
Port | Connected Component |
|---|---|
1 | Viper plug |
2 | Waste |
3 | Injection valve |
4 | Viper plug |
5 | Check valve |
6 | Pressure sensor |
7 | Waste |