Pump head and components
Description
Part No.
Pump head (complete assembly)
6251.1102
Piston
6251.1155
Seal-handling tool
6251.5122
Piston seal (RP) and seal wash seal, pump head (pack of 2 each)
6251.1143
Support ring (pack of 2)
6040.0012
Check valve, inlet check valve
6251.1153
Check valve, outlet check valve
6044.2310
Capillary kit, pump head, including:
  • U-tube
  • Capillary from pump head to flowmeter
6251.0086
VN rear sealing and screws kit (VN pump), seal wash plate/pump head, including:
  • Pump head seal, for seal wash plate (O-ring seal, 11x1.5) (2 seals)
  • Pump head seal, for seal wash plate screw (O-ring seal, 6.5x1) (8 seals)
  • Screw, seal wash plate (8 screws)
6251.2234
Flowmeter
Description
Part No.
Filter kit, flowmeter, including filter screw, screw seal, and filter frit (2 µm)
To replace the filter or filter frit on both flowmeter inlets, two kits are needed.
6251.0111
Flowmeter waste line, see Low-pressure tubing kit
Low-pressure tubing kit
Description
Part No.
Low-pressure tubing kit, pump, including:
  • Flowmeter waste line from flowmeter to waste outlet port
  • Solvent lines (A, B) from shut-off valves to pump heads
  • Solvent lines from degasser to shut-off valve
6251.0150
Also see Solvents and Wash Systems and Capillaries, Capillary Accessories and Drainage.
Maintenance kit
Description
Part No.
Pump maintenance kit, including:
  • Screws, seal wash plate (8 screws)
  • Silicone tubing (clear tubing)
  • Peristaltic tubing (PharMed, white tubing)
  • Tubing connector (straight)
  • Tubing connector (90°-angled)
  • Tubing connector (I.D. 1/16") (pack of 4)
  • Piston seal (pack of 4)
  • Pump head seal, for seal wash plate (O-ring seal, 11x1.5)
    (pack of 2)
  • Pump head seal, for seal wash plate screws (O-ring seal, 6.5x1)
    (pack of 8)
  • Seal wash seal (pack of 4)
  • Solvent line filter frit (biocompatible, 10 µm) (pack of 10)
6251.0130
Miscellaneous
Description
Part No.
NanoViper capillary, fused silica, I.D. x length 20 µm x 350 mm,
for use as backpressure capillary during pump maintenance
6250.5240
Fitting plug, Viper, biocompatible
6040.2303
Syringe and tubing kit, including
  • 12 mL plastic syringe (pack of 5)
  • Silicone tubing (O.D. x I.D. 2.8 x 1.3), 3 m
6000.0010
Rinsing tubing, silicone (O.D. x I.D 2.8 x 1.3), 3 m
6007.9100
Cleaning swabs (pack of 25)
6040.0007
Fuses kit, Vanquish system
For the pump, autosampler, and column compartment, use the 5 AT, 230 V AC, slow-blow fuses.
6036.0002