Follow these steps
  1. Take the cover by the bottom end and slightly pull the cover to release the cover groove from the insulation cover mounting bracket.
  2. Carefully pull the cover to release it from the top enclosure frame.
  3. Pull the cover away from the insulation cover mounting bracket.
Figure: Removing the insulation cover