When
  • To install replacement pump head seals after seal removal
  • To reinstall a pump head seal that came off during maintenance of pump head parts
Parts and tools required
  • Pump head seals
  • Screwdriver, Torx T30
  • Syringe filled with isopropanol
  • Lint-free tissue
Parts affected on the pump head
123456
Figure: Pump head seals (here: left side of pump head)
No.
Description
1
Pump head
2
Pump head seals, for seal wash plate screws (O-ring seals 6.5x1)
3
Pump head seal, for seal wash plate (O-ring seal 11x1.5)
4
Seal wash plate
5
Seal wash plate screws
6
Piston
Follow these steps
  1. Only when reinstalling a pump head seal: Clean the pump head seal, using isopropanol and a lint-free tissue.
  2. Place the pump head seal onto the receiving groove.
11112
Figure: Receiving grooves for pump head seals (here: left side of pump head), in pump head and seal wash plate
No.
Description
1
Receiving grooves for pump head seals in pump head; seals for seal wash plate screws (O-ring seals 6.5x1)
2
Receiving groove for pump head seal in seal wash plate (side facing the pump head); seal for seal wash plate (O-ring seal 11x1.5)
  1. With your fingers, press the pump head seal into the groove as far as it goes in.
Figure: Pump head seals installed, in pump head and seal wash plate (here: left side of pump head)
  1. Place the seal wash plate onto the pump head and insert the screws.
  2. Figure: Screws on seal wash plate (here: left plate)
  3. With the screwdriver, tighten the screws crosswise.
  4. Tighten the screws crosswise once again.
  1. Install the piston. See Installing the Pistons.
  2. Install the pump head. See Installing the Pump Head.