Parts required
Insulation cover
Follow these steps
  1. Properly route the sample loop capillary through the designated guide in the insulation cover mounting bracket.
  2. Figure: Insulation cover with mounting bracket (left); mounting bracket with sample loop capillary guide (right)
  3. Align the cover with the needle unit and the sample loop on the interior front panel.
  4. Carefully push the cover groove on the insulation cover mounting bracket to insert the insulation cover. Do not pinch the sample loop capillary with the cover.