When
To install a replacement seal wash seal after seal removal
Parts and tools required
  • Seal wash seal
  • Seal-handling tool
12123
Figure: Seal-handling tool (left: closed tool; right: open tool)
No.
Description
1
Removal pin
2
Insertion pin
3
Knurl for opening and closing the seal-handling tool
(opening and closing is required only when using the removal pin)
Parts affected on the pump head
1342
Figure: Seal wash seal (here: for left piston)
No.
Description
1
Pump head
2
Seal wash plate
3
Seal wash seal
4
Piston
  1. Slide the seal wash seal onto the insertion pin of the seal-handling tool. Mind the orientation of the seal.
1221
Figure: Sliding the seal wash seal onto the insertion pin
No.
Description
1
Seal wash seal (the arrow indicates which side faces the tool)
2
Insertion pin, seal-handling tool
  1. With the insertion pin, push the seal-handling tool into the seal wash plate and remove the tool. The seal wash seal remains in the seal wash plate.
  2. Figure: Seal wash seal installed (here: for left piston)
  3. Install the piston. See Installing the Pistons.
  4. Install the pump head. See Installing the Pump Head.