When
Damage of seal wash seal, liquid observed underneath the pump head (leaky seal)
Follow these steps
Remove the seal wash seal. See Removing the Seal Wash Seal.
Install the seal wash seal. See Installing the Seal Wash Seal.
When
Damage of seal wash seal, liquid observed underneath the pump head (leaky seal)
Follow these steps
Remove the seal wash seal. See Removing the Seal Wash Seal.
Install the seal wash seal. See Installing the Seal Wash Seal.