Parts required
  • LAN cable
  • Network card
Follow these steps
NOTICE
Vanquish module damage
The System Interlink and the LAN ports use the same connector type. Connecting a LAN cable, for example, to a System Interlink port of a Vanquish module, may damage the Vanquish module and the system controller.
Only connect matching ports.
  1. Connect a LAN cable to the LAN port of the system controller.
  2. Connect the other connector of the LAN cable to the network card LAN port of the computer with the chromatography data system.