When
  • To replace a piston
  • For maintenance of pump head parts
Preparations
Remove the pump head. See Removing the Pump Head.
Follow these steps
With your fingers, pull out the piston that you want to remove from the pump head. If you remove the piston during maintenance of pump head parts, grasp the piston by the piston base and avoid touching the piston surface.
Figure: Removing the pistons