CAUTION
Heavy Load, Bulky Device
The system modules are too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the system modules, observe the following guidelines:
Tools required
Screwdriver, Torx T20
Preparations
Follow these steps
Loosen the four captive screws on the front left and front right of the device.
Push all tubing and capillaries, which are present in the tubing chase of the Vanquish system modules, into the tubing chase.
Grasp the slide-in module by the leak tray, and pull the module out of the enclosure approximately 10 cm.
NOTICE
The slide-in module can fall down when pulling it out of the enclosure too far.
Remove the slide-in module from the enclosure. The following steps require a team effort:
- Take the slide-in module on both sides from below.
- Pull the slide-in module from the rails towards the front.
- Place the slide-in module on a clean and stable surface.