Capillaries
Description
Part No.
Capillary pump-autosampler, nanoViper, fused silica,
I.D. x length 20 µm x 350 mm
6250.5240
Capillary autosampler-column compartment/column, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x length 20 µm x 550 mm
6250.5260
Capillary pump-autosampler, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x length 50 µm x 350 mm
6250.5540
Capillary autosampler-column compartment/column, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x length 50 µm x 550 mm
6250.5560
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 20 µm x 280 µm x 1 m, for nano LC applications
6041.5292
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 20 µm x 360 µm x 1 m, for nano LC applications
6041.5293
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 50 µm x 280 µm x 1 m, for nano LC applications
6041.5294
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 50 µm x 360 µm x 1 m, for nano LC applications
6041.5295
Capillary accessories
Description
Part No.
Fitting plug, Viper, biocompatible
6040.2303
Viper union
6040.2304
Capillary tools
Description
Part No.
Tubing cutter for fused silica tubing
6720.0016
Tubing cutter for PEEK tubing
6300.0401
Torque screwdriver, Viper, base part with extension adapter and bits for tightening nanoViper capillaries and the autosampler needle seat
6250.2110
Drainage kit
Description
Part No.
Drainage kit, Vanquish system, including:
  • L piece (elbow, pack of 4, different sizes)
  • Drain tubing, 6.25 m length
  • Drain tubing clips (pack of 2)
6036.1120