The system comprises the following features:
Main Features
  • Pre-installed system offering high workflow versatility
  • Operating pressure of up to 150 MPa
  • Flow range from 100 nL/min to 100 µL/min
  • Integrated automated startup and purging functions
  • Continuous background system monitoring for minimized unplanned downtime
  • Diagnostic features for fast troubleshooting and increased uptime
  • Touch-based user interface, offering instrument monitoring and control and maintenance assistance
Specific Pump Features
  • Binary pump with each channel based on a serial dual-piston design
  • Floating piston design for compensation of small mechanical tolerances within the specification and thus enhanced robustness of the pump
  • One flowmeter supporting flow rates of 100 nL/min to 100 µL/min;
    no hardware change required except for system capillaries
  • Active seal wash system for high lifetime of pistons and piston seals
  • Automated purge valves for easy startup and diagnostics
  • Integrated automated startup and purging functions
  • Built-in automatic shut-off valves preventing solvent spillage and ensuring robust solvent aspiration
  • Built-in vacuum degasser for autosampler needle wash liquids
Specific Autosampler Features
  • Low carry-over split-loop injection principle featuring two needle wash liquids (weak and strong)
  • Washing inner needle, needle seat, and sample loop with weak and strong wash liquid
  • Washing outer needle surface with weak and strong wash liquid for minimum LCMS carry-over
  • Injection volumes up to 100 µL with one single stroke
  • Injection volumes higher than 100 µL through multi-draw feature for preconcentration of large volumes
  • Integrated automated startup and purging functions
  • Bottom sensing to allow aspiration of small volumes with minimal sample loss
  • Temperature control for thermostatting the sample compartment
  • Barcode reader inside the autosampler for identification of sample racks and well plates with barcodes for Vanquish rack type identification
  • Support of sample racks and well plates with a footprint as specified by the Society for Biomolecular Screening (SBS footprint)
  • Seal wash for metering device head for high lifetime of the seals
  • Integrated loading functionality
Specific Column Compartment Features
  • Column chamber (34 x 8 x 4 cm, w x h x d) providing room for columns, trap columns, and capillaries in various workflow setups
  • Space for up to two switching valves inside the column chamber for advanced applications
  • The column chamber and the components inside the chamber can be thermostatted over a wide temperature range with the minimum temperature being 5 °C above ambient temperature and the maximum temperature being 80 °C without valves or 60 °C with Vanquish Neo low-dispersion (LD) valves
  • Optimized design enabling an even air flow and heat distribution
  • Fast heat-up time (35 °C to 65 °C ±1 °C in less than 12 minutes)
  • Advanced temperature control providing temperature stability of ±0.05 °C