Inside the column chamber, eight column brackets are installed. Attach the columns to the column brackets using the column clips from the ship kit. You can install the column clips on the column brackets in any position.
Figure: Possible column clip positions
Parts required
  • Column as required by the application
  • Column clips
  • Self-adhesive foam pads
Follow these steps
  1. To remove the ring from the bottom part of the column clip, slightly press the ring in the direction of the arrows.
  2. 112323
    Figure: Column clips
    No.
    Description
    1
    Ring
    2
    Foam pad
    3
    Bottom part
  3. Self-adhesive foam pads are shipped together with the column clips. To protect the column, attach one pad to the inside of each bottom part.
  4. Attach the bottom part of the column clip to the column bracket in the desired position and turn it 90° clockwise.
  5. Connect the capillaries to the column inlet and column outlet.
  6. Press the column into the bottom part of the column clip and reinstall the ring.
  7. Figure: Reinstalling the ring
  8. Route the capillaries through one of the capillary guides shown in the figure.
  9. Figure: Capillary guides
  10. To set the column properties, run the scripts: A03 - Set Separation Column Type and A04 - Set Separation Column Specifications (and A05 - Set Trap Column Specifications, as required).
  11. Turn on the pump flow.