Damaged Packaging, Defective on Arrival
Inspect the shipping container for signs of external damage and, after unpacking, inspect the content for any signs of mechanical damage that might have occurred during shipment.
If you suspect that the content may have been damaged during shipment, immediately notify the incoming carrier and Thermo Fisher Scientific about the damage. Shipping insurance will compensate for the damage only if reported immediately.
Unpacking
NOTICE
To lift or move the device, grasp the device by the carrying handles. Do not move or lift the device by the front doors. This will damage the doors or the device.
Follow these steps
Place the shipping container on the floor and open it.
Remove the ship kit from the shipping container.
Remove the device from the shipping container: Grasp the device by the carrying handles. Slowly and carefully, lift the device out of the shipping container.
Place the device on a stable surface.
Remove any additional packing material. Some surfaces including the doors of the device are covered by a protective film during shipment. Leave any protective films attached to the surfaces of the device until it is properly positioned in the system stack.
Transport the device by the carrying handles to the installation site, if it is not already there, and place it in the system stack. See Placing the Column Compartment in the System Stack.
Remove the protective films from all surfaces as applicable.