When
System needs to be transported to another location.
Parts required
  • 4 handles
    Use the carrying handles that were installed when the system was shipped and that have 2 holes for screws.
  • Figure: Carrying handle for the system stack
  • 8 screws that were fixed to the handles
Tools required
Screwdriver, Torx T30
Follow these steps
CAUTION
Personal injury and system damage
Fixing the handles to the autosampler and lifting the system could lead to personal injury and to system damage.
If you need to lift and carry the whole system, fix the handles only to the pump, to avoid personal injury and system damage.
  1. Insert a carrying handle into the right and into the left rail from the rear.
  2. Figure: Sliding the carrying handle on the right rail
  3. Slide the handles toward the system's front until the distance between the front of the rail and the front edge of the handle is about 6 cm.
  4. 6 cm6 cm
    Figure: Positioning the front carrying handles (upper image: stack seen from the left; lower image: stack seen from the right)
  5. Fix the screws of the handles.
  6. Insert a carrying handle into the right and into the left rail from the rear.
  7. Slide the handles toward the system front so that the rear side of the handles flushes with the rear side of the rails.
  8. Figure: Correct position of the rear carrying handles
  9. Fix the screws of the handles.
CAUTION
Risk of Injury and Module Damage
Handles can slide off the module if the attachment screws are not fixed properly. Also, a slide-in module can fall out if not fixed properly. This could cause personal injury and damage to the module. Verify the following before lifting the system:
  • The handles are fixed properly and cannot slide off the module.
  • The four screws of each slide-in modules are tightened and no slide-in module can fall out (see Replacing the Slide-In Module).