Tools required
Screwdriver, Torx T10
Follow these steps
If personnel other than a Thermo Fisher Scientific service engineer installs the system, follow the steps below.
  1. To be able to shift the system base slightly on the workbench for better access to system parts, unlock the system base using the system base locking tools. See Unlocking the System Base.
  2. Move the system stack into an appropriate working position.
  3. Unplug the two display connectors from the system controller.
  4. Remove the cable cover.
  5. Remove the display cable from the rail and the housing cable clips.
  6. Holding the fastening block with the right hand, unfasten the screw in one of the holes marked with a circle with the left hand from the outside.
  7. Figure: Unfastening the screw holding the fastening block
  8. Remove the fastening block.
  9. Unfasten the 2 screws in the two upper holes.
  10. Figure: Unfastening the screws
  11. Push the display assembly toward the system's rear and remove it from the stack.
  12. Fasten the upper 2 screws and fasten the fastening block with a screw.
Figure: Screws to be fastened on the display assembly