You can find the user-accessible components and flow connections in the devices directly behind the doors.
Opening the Front Doors
  1. Open the left door.
  2. Open the right door.
Opening Mechanism of the Autosampler
The left front door of the autosampler is equipped with a mechanism that opens the door automatically to ensure proper ventilation of the sample compartment when the autosampler is turned off. The door also opens automatically if a power failure occurs or if you disconnect the power cord while the autosampler has been turned on.
Opening the front doors (here: autosampler)
Figure: Opening the front doors (here: autosampler)
Closing the Front Doors
  1. Close the right door.
  2. Close the left door.
Closing Mechanism of the Autosampler
When the autosampler is turned on, the left front door can be closed completely.