A Thermo Fisher Scientific service engineer installs and sets up the Vanquish Neo system, including all modules and options or parts shipped with them. The service engineer checks that the installation is correct and that the Vanquish Neo system and modules operate as specified. The engineer also demonstrates the basic operation and main features.
If personnel other than a Thermo Fisher Scientific service engineer installs the system, follow the steps below.
The steps in the diagram are basic installation steps. Follow the in-depth instructions of each step in the diagram in the given sequence.
Observe the safety guidelines |
↓ |
Observe all site requirements |
↓ |
Optional: Install the column compartment and accessories |
↓ |
Remove the transport protections from the autosampler |
↓ |
Mount the display |
↓ |
Connect cables |
↓ |
Optional: Connect a mass spectrometer |
↓ |
Set up the flow connections |
↓ |
Fill the liquid reservoirs |
↓ |
Turn on the system |
↓ |
Integrate the system into an instrument LAN |
↓ |
Prepare the system for initial operation |