In the autosampler, you can thermostat the sample compartment.
Follow these steps
Enable temperature control in the chromatography data system or in the autosampler box on the home panel in the Vanquish User Interface.
Set the target temperature. Check that the thermostatting temperature is suitable for the samples.
TIP
If you turn off the autosampler, the autosampler saves the set target temperature. Check the temperature when you turn the autosampler on again and adapt it if required.