The figure shows the parts of the solvent rack.
2c2a1a2b12
Figure: Solvent rack parts
No.
Description
1
Solvent rack
1a
Magnetic front
Allows attaching a magnetic label holder to the front side of the solvent rack, for example, to show a specific system name. Labels are included in the system ship kit.
2
Bottle insert1
Provides slots for the following reservoirs:
2a
5 slots for 1 L reservoirs
2b
2 slots for 2 L reservoirs
2c
4 slots for 0.25 L reservoirs
1: The bottle insert can be removed easily. If you remove the bottle insert, preferably use only the liquid reservoirs that are shipped with the system. If you use other liquid reservoirs, liquid reservoirs with more capacity, for example, ensure that the system stack is stable and does not tilt. If you doubt safety, install a stack stabilization kit (see Ship Kit).