The system interlink connections require a special system interlink cable (Cat. 6 cable).
System interlink cable
Figure: System interlink cable
The system interlink connections from one system module to the other are pre-installed except for the optional column compartment.
1234
Figure: System interlink cable connections
No.
Description
-
System interlink connection:
1
From system base to system controller
2
From system controller to pump
3
From pump to autosampler
4
From autosampler to column compartment (if installed)
If a Column Compartment is Installed (optional)
  1. Remove the cap which covers the right System Interlink port on the autosampler.
  2. Connect one end of the system interlink cable to the System Interlink port on the column compartment, and the other end to the right System Interlink port on the autosampler (as shown in the figure).