The user-accessible components of the column compartment are located directly behind the front doors.
123457xxxxxxxxxxxxxxxxxxxxxxxx866669910101010
Figure: Column compartment interior components
No.
Description
1
Keypad
2
Left column switching valve (if installed; here: Vanquish valve)
3
Column ID ports
4
Fan
5
Temperature sensor
6
Capillary guides
7
Right column switching valve (if installed; here: Vanquish valve)
8
Type label, indicating the module name, serial number, part number, and revision number
9
Mounting positions for the passive pre-heater
10
Column brackets