The digital I/O ports (Dig I/O) can be used to exchange digital signals with external instruments, for example, a mass spectrometer. Each port provides:
  • one digital input
  • one relay output
Digital I/O port
Figure: Digital I/O port
Pin
Description — Signal Name
1
Not used
2
Relay output — Relay_NC (Normally Closed contact)
3
Ground — GND
4
Digital input — Input
5
Relay output — Relay_COM
COM is the common contact for NO and NC. If the relay is not activated or if the device is turned off, the connection is between COM and NC. If the relay is activated, the connection is between COM and NO.
6
Relay output — Relay_NO (Normally Open contact)
The next table lists the functions assigned to the connector pins and the color of the cable wire connected to each pin.
Pin
Wire Color
Signal Name
Signal Level
Remarks
1
Pink
Not used
2
Gray
Relay output — Relay_NC
Potential free
0-24 V, 0-100 mA
Opening contact
3
Green
Ground — GND
Ground
Reference potential
4
Yellow
Digital input — Input
Input (low active):
On: 0-0.4 V
Off: 2.2-5 V
Pull-up resistor: 47 kΩ to 5 V
Digital input; reference potential is ground. Note the following:
  • The maximum input voltage at the input must not exceed +5 V with reference to ground.
  • The minimum input voltage must not be lower than the ground potential.
5
White
Relay output — Relay_COM
Potential free
Common contact for NO and NC
6
Brown
Relay output — Relay_NO
Potential free
0-24 V, 0-100 mA
Closing contact