The pump performance is specified as follows:
Type
Specification
Operating principle
Serial dual-piston pump with independent piston drives
Compressibility compensation
Fully automated, independent of the composition of the mobile phase
Flow range (settable)
nL/min – 100 µL/min in 1 nL/min increments
Flow accuracy
Typically: ± 3%
Conditions: 0.1 - 100 µL/min any of 4 pre-calibrated solvents, measured at 20 MPa system pressure
Inter-module deviation
< 5% for flow rates > 300 nL/min – 10 µL/min
Conditions: Measured by retention time differences of TGQAPGFSYTDANK Cytochrome C protein digest peptide and 30 min gradient.
Retention time precision
≤ 0.2% RSD or ≤ 0.1 SD min, whichever is greater
Conditions: For TGQAPGFSYTDANK Cytochrome C protein digest peptide for gradients 30 – 90 min.
Pressure range
Pump pressure: 2 – 150 MPa (20 – 1500 bar, 300 – 21750 psi)
150 MPa pump pressure up to 1 µL/min. With a flow rate above 1 µL/min, the maximum pressure is solvent dependent and decreases linearly down to 70 MPa at 100 µL/min for acetonitrile 80% in channel B.
Number of solvent channels
2
Maximum stroke volume
96 µL
Dwell volume
(contribution of the pump to the system gradient delay volume)
25 nL
Degassing
2 channels for autosampler wash liquids
Communication
USB:
1 USB port (USB 2.0, "B" type connector)
1 USB hub with 3 ports (USB 2.0, "A" type connectors)
I/O Interface:
2 Dig I/O ports (mini-DIN), each providing one input and one relay output
System Interlink:
2 System Interlink ports (RJ45-8 connectors)
Materials in the analytical flow path
Ceramics, DLC (diamond like carbon), fluoropolymers, MP35N, PEEK, titanium, UHMW PE
For information on the chemical resistance of materials, refer to the technical literature.