The column compartment performance is specified as follows:
Type
Specification
Pressure range
Vanquish valve: 50-1500 bar
VICI valve C82: 50-1034 bar
VICI valve C2, C2N: 50-345 bar
Temperature range
Min.: 5 K above room temperature
Max.: 80 °C without valves installed
Max.: 60 °C with Vanquish valves installed
Max.: 50 °C with VICI valves installed
Temperature accuracy
±0.5 °C
Temperature stability
±0.05 °C
Temperature precision
±0.1 °C
Heat up time
From 35 °C to 65 °C ±1 °C in less than 12 minutes
Column switching valves
Up to two valves:
2-position 6-port Vanquish valve, 1500 bar
2-position 6-port C82 VICI valve, 1034 bar
2-position 6-port C2 VICI valve, 345 bar
2-position 6-port C2N VICI valve, 345 bar
For further information on available column switching valves, see Optional Accessories.
USB communication
1 USB port (USB 2.0, "B" type connector)
1 USB hub with 3 ports (USB 2.0, "A" type connectors)
I / O Interface
2 Dig I/O ports (mini DIN), each providing one digital input, and one relay output
System Interlink
2 System Interlink ports (RJ45-8 connectors)
Materials in the flow path
Vanquish valve: ceramic, DLC, titanium
VICI valve C82: stainless steel with inert coating, PAEK composit
VICI valve C2, C2N: PAEK, PAEK composit
Capillaries: MP35N (for details, see the pre-heater specifications)
For information about the chemical resistance of materials refer to the technical literature.
Passive pre-heater
Type
Specification
Temperature range
Same as column chamber
Wetted parts
MP35N
Biocompatible
Yes
Heated volume
1 µL
Total volume
6 µL