3b212a2b2d2e3a3c3d3e3f43g33h2c
Figure: Operating principle
No.
Description
1
Solvent lines: A and B
2
Flow-controlled pump with two identical pump blocks (solvent channels), A and B
2a
Pump blocks A and B, each pump block with:
2b
Working cylinder
2c
Working pressure sensor
2d
Equilibration cylinder
2e
Primary pressure sensor
The primary pressure is the pressure in the pump head.
3
Flowmeter, as applicable for each channel, with:
3a
Flowmeter inlet line (capillary from pump head)
3b
Flowmeter filter
3c
Automatic purge valve with waste outlet
3d
Flow sensor
3e
Restrictor capillary
3f
System pressure sensor
3g
Flowmeter outlet with fluidic Tee piece (pump outlet)
3h
Waste outlet
4
Capillary to system
The pump is a binary pump with a flowmeter for flow control:
  • Each pump block has a working cylinder and an equilibration cylinder. The cylinders are connected in series. The solvent passes the cylinders successively.
  • Each pump block delivers flow continuously.
    The working cylinder delivers while the equilibration cylinder is filled simultaneously. The equilibration cylinder serves as the reservoir and delivers while the working cylinder performs the intake stroke.
  • The sensors in the flowmeter measure and control the partial flows of the pump blocks.
  • The partial flows are combined in the fluidic Tee piece at the flowmeter outlet. The combined flow leaves the flowmeter with the selected target flow rate and solvent composition.