The table lists the safety symbols that appear on the system or on labels affixed to the system. Follow the safety notices in this manual to prevent the risk of operator injury or damage to the system.
Symbol | Description |
|---|---|
| Indicates a potential hazard. Refer to this manual to avoid the risk of personal injury and/or to prevent damage to the device. |
— Ο | Power supply is on Power supply is off |
Indicates a power button that switches the system between on and off states. | |
Indicates a power button that switches the system controller on and off. | |
Indicates alternating current. | |
| Indicates that the surface becomes hot during operation. Do not touch these surfaces while they are heated up. |
| Indicates a potential pinch point hazard. Keep your hands clear to avoid harming your hands. |
Indicates a terminal for protective grounding Can be used to attach a device for protective grounding, for example, a column. |