Allowed concentrations for the system:
Concentration | Value |
|---|---|
Buffer concentration | n.a. |
Chloride concentration | Less than or equal to 1 mol/L High chloride concentration: The application time should be as short as possible. Flush the system thoroughly after these applications. |