Allowed pH range for the system:
Allowed pH range | Remark |
|---|---|
2-10 | pH values of 2: The application time should be as short as possible. Flush the system thoroughly after these applications. |
Allowed pH range for the system:
Allowed pH range | Remark |
|---|---|
2-10 | pH values of 2: The application time should be as short as possible. Flush the system thoroughly after these applications. |