Tools required
Scissors for cutting the cable cover
Follow these steps
Plug the display power cable into the Display DC OUT 24V port of the system controller.
Fasten in the display data cable to the Display Data port of the system controller finger-tight.
Route the display power and data cables in the rail.
Cut the cable cover to a length of 34 cm with the scissors.
Firmly push the cable cover into the rail.
Fix the vertical part of the cable route with the housing cable clips.
Move the system stack back into the operating position.
Lock the system base using the system base locking tools (see Locking the System Base).