You can fix the display to the left side of any system stack module (if the module meets the requirement mentioned under Prerequisite). To swivel the display, keep a clearance of 10 cm to the left.
Prerequisite
You can fix the display only to modules that have housings with an angular notch. The display is not suitable for housings with a U-type notch.
Figure: Housings with notches (left: angular, right: U-type)
Preparations
Unpack the display.
TIP
Keep the shipping container and all packing material. These items will be needed if the display is transported to a new location.
Parts required
  • Display assembly including:
    • 3 Torxâ„¢ screws
    • Fastening block
    • Sliding block
  • 12
    Figure: Sliding and fastening blocks
    No.
    Description
    1
    Sliding block
    2
    Fastening block
  • Display stop
  • Cable cover
Tools required
Screwdriver, Torx T10
Follow these steps
If personnel other than a Thermo Fisher Scientific service engineer installs the system, follow the steps below.
  1. Check whether the display cable is as shown below.
  2. 12
    Figure: Correct orientation of the display cable
    No.
    Description
    1
    Swivel arm
    2
    Display cable
  3. If the display cable is not as shown, turn the display clockwise or counterclockwise at the swivel arm until the display cable is guided as shown above.
  4. Figure: Turning the display at the swivel arm
  5. Loosen the upper 2 screws and unscrew the lower screw on the display assembly to remove the fastening block.
  6. Figure: Affected screws on the display assembly
  7. From the rear side of the module rail, insert the sliding block of the display into the rail on the left side of the module and pull the assembly toward the system front until it reaches the stop.
  8. 213
    Figure: Sliding in the display
    No.
    Description
    1
    Display assembly
    2
    Sliding block
    3
    Rail
  9. Slightly fasten the display assembly with 2 screws in the two upper holes.
  10. Figure: Fastening the display
  11. On the inside of the left housing panel, locate the notch.
  12. Figure: Notch
  13. Place the fastening block at the height of the notch as shown. Make sure that the hole in the fastening block matches that of the housing.
  14. Figure: Placing the fastening block
  15. Holding the fastening block with the right hand, slightly fix it with a screw with the left hand on the outside. Use one of the holes marked with a circle in the image below.
  16. Figure: Fixing the fastening block
  17. Check whether the display is in a horizontal position. If not, align it as follows:
  1. Move the display slightly until it is in a horizontal position.
  2. Firmly fasten the 3 screws.
  1. If the display is attached to the top module, place the display stop to the solvent rack as shown.
Figure: Display stop