A barcode reader inside the sample compartment allows automatic identification of the rack type for sample racks and well plates on which a Vanquish rack type 2D barcode is present.
During operation, the barcode reader performs an inventory scan and reads the Vanquish rack type barcode if present. The barcode reader automatically identifies the rack type and orientation. This information is sent to the chromatography data system.
To allow the identification of the sample rack or well plate type, use sample racks and well plates with such barcodes for rack type identification.
Each segment accommodates a barcode label that informs the barcode reader that the segment is empty if no sample rack or well plate is installed.
12
Figure: Sample rack with barcodes for rack type identification
No.
Description
1
Sample position A1
2
Vanquish rack type barcode