The carousel separates into four color-coded segments: red (R), green (G), blue (B) and yellow (Y).
Each segment separates into the following areas:
  • Positioning area that accommodates space for one sample rack or well plate
  • Support vial holder that comprises three vial positions for vials with a volume of up to 10 mL, such as reagent vials
    The support vial holder is positioned left of the area for the sample rack or well plate in each segment.
Detailed view on segment in the carousel
Figure: Detailed view on segment in the carousel
No.
Description
1
Barcode label 'empty'
Label that identifies that the segment is empty for the barcode reader
2
Color code for the segment (here: R for the red segment)
3
Positioning area of the segment
Space to position a sample rack or well plate
With alignment angles and alignment points to align the sample rack or well plate
4
Support vial holder
For three vials with a volume of up to 10 mL
5
Color code for the support vial holder (here: R1 for example for the first vial holder in the red segment)