The sample compartment of the autosampler accommodates a carousel with four color-coded segments, each with an additional three-position vial holder for 10 mL vials.
The autosampler supports sample racks and well plates with a footprint as specified by the Society for Biomolecular Screening (SBS footprint).
The sample compartment provides the following features:
Sample compartment thermostatting, see Sample Compartment Thermostatting.
Rack type identification and verification, empty segment detection and inventory management, see Rack Type Identification.