When
To empty the solvent lines, for example, for maintenance or service procedures
Items required
Solvent suitable to flush out harmful substances if necessary
Follow these steps
  1. To flush out harmful substances, if applicable, purge the pump with an appropriate solvent.
  1. Unscrew the cap of the solvent reservoir and remove the cap together with the solvent line from the reservoir.
  2. Start a purge cycle as appropriate.
  3. Stop purging as soon as the solvent line is empty.