The picture shows the main pump head parts.
12345678910111213
Figure: Overview of pump head parts
No.
Description
1
Pump head
2
Inlet check valve
3
Piston seals
4
Support rings
5
Pump head seals, for seal wash plate screws (O-ring seals 6.5x1)
6
Pump head seals, for seal wash plates (O-ring seals 11x1.5)
7
Seal wash plates
8
Screws, seal wash plates
9
Seal wash seals
10
Pistons
11
Connectors for seal wash tubing (screwable)
12
Outlet check valve
13
U-tube