When
Contamination of detector electrodes
Impaired functionality of the seal wash system
Parts required
Seal wash detector assembly, consisting of the detector and funnel
Preparations
To prepare the new seal wash detector assembly push the seal wash detector into the funnel. Mind the orientation of the funnel.
No. | Description |
|---|---|
1a | Funnel (bottom side) |
1b | Funnel (top side) |
2 | Seal wash detector |
Follow these steps
No. | Description |
|---|---|
1 | Funnel, seal wash detector |
2 | Seal wash detector |
3 | Detector cable |
4 | Seal wash line |
Disconnect the seal wash line from the seal wash detector (port labeled in).
Unplug the detector cable from the DROP DET port.
Grasp and pull the seal wash detector assembly upward by the funnel.
Push the new seal wash detector assembly into the pump.
Reconnect the seal wash line to the seal wash detector (port labeled in).
Connect the detector cable to the DROP DET port.
Flush the seal wash system. Run the script: B01 - Change Liquids / Solvents (with: Rear Seal Wash Liquid and Refresh Only = Off.)