If you want to change the seal wash liquid:
Run the script: B01 - Change Liquids / Solvents (with the following parameters: Rear Seal Wash Liquid and Refresh Only = Off).
If you want to change the seal wash liquid:
Run the script: B01 - Change Liquids / Solvents (with the following parameters: Rear Seal Wash Liquid and Refresh Only = Off).